missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbonylhydridotris(triphenylphosphine)iridium(I), TCI America™

Manufacturer:  TCI America C30401G

 View more versions of this product

Catalog No. C30401G



Specifications

Specifications

Carbonylhydridotris(triphenylphosphine)iridium(I)
170°C
17250-25-8
MFCD00015525
carbonylhydridotris triphenylphosphine iridium i, iridium 1+ formaldehyde tris triphenylphosphine hydride
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ir]
1010.126
1008.11
Yellow
1g
C55H48IrOP3-
2811
NTACLHNBUTVRBR-UHFFFAOYSA-N
formaldehyde;hydride;iridium;triphenylphosphane
92135237
Crystal-Powder at 20°C

Chemical Identifiers

17250-25-8
1010.126
NTACLHNBUTVRBR-UHFFFAOYSA-N
92135237
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Ir]
C55H48IrOP3-
MFCD00015525
carbonylhydridotris triphenylphosphine iridium i, iridium 1+ formaldehyde tris triphenylphosphine hydride
formaldehyde;hydride;iridium;triphenylphosphane
Safety and Handling

Safety and Handling

TSCA : Yes

Documents