missing translation for 'onlineSavingsMsg'
Learn More

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

CAS: 16971-33-8 | C55H46ClOP3Ru | 952.41 g/mol

$181.18 - $181.18

Chemical Identifiers

CAS 16971-33-8
Molecular Formula C55H46ClOP3Ru
Molecular Weight (g/mol) 952.41
MDL Number MFCD00049804
InChI Key IMPHYUNMDWWJAH-UHFFFAOYSA-M
Synonym carbonylchlorohydridotris triphenylphosphine-ruthenium ii
PubChem CID 71463878
IUPAC Name formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
SMILES Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC392140010
SDS View Documents
Thermo Scientific Chemicals
392140010
1 g Glass bottle
Each for $181.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

16971-33-8
952.41
IMPHYUNMDWWJAH-UHFFFAOYSA-M
71463878
Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C55H46ClOP3Ru
MFCD00049804
carbonylchlorohydridotris triphenylphosphine-ruthenium ii
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride

Specifications

16971-33-8
Cream to Tan
C55H46ClOP3Ru
1 g
Solubility in water: insoluble
Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
952.41
952.4
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
204.0°C to 206.0°C
Glass bottle
MFCD00049804
carbonylchlorohydridotris triphenylphosphine-ruthenium ii
IMPHYUNMDWWJAH-UHFFFAOYSA-M
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
71463878
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-051-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only