missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Manufacturer:  Alfa Aesar™ 4495503

 View more versions of this product

Catalog No. AA4495503



Specifications

Specifications

Carbonylchlorohydridotris (triphenylphosphine)ruthenium (II)
Gray
1g
C55H48ClOP3Ru
MFCD00049804
ITDZWCWUTIWRCJ-UHFFFAOYSA-M
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
71463878
Crystalline
Moisture & Light Sensitive
204°C to 206°C (decomposition)
16971-33-8
[(C6H5)3P]3Ru(CO)(Cl)H
carbonylchlorohydridotris triphenylphosphine-ruthenium ii
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ru+2]
954.429
952.41

Chemical Identifiers

16971-33-8
954.429
ITDZWCWUTIWRCJ-UHFFFAOYSA-M
71463878
[H-].C=O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Ru+2]
C55H48ClOP3Ru
MFCD00049804
carbonylchlorohydridotris triphenylphosphine-ruthenium ii
formaldehyde;hydride;ruthenium(2+);triphenylphosphane;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents