Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonylchlorobis(triphenylphosphine)rhodium(I), Rh 14.9% min

Chemical Identifiers

Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)
CAS 13938-94-8
Linear Formula RhCl(CO)[P(C6H5)3]2
Formula Weight 690.94
MDL Number MFCD00044273
Beilstein 4618896
Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3982201

  SDS
alfa aesar™
3982201
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982203

  SDS
alfa aesar™
3982203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3982206

  SDS
alfa aesar™
3982206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 690.94
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)rhodium(I)
Odor Odorless
Physical Form Crystalline
Linear Formula RhCl(CO)[P(C6H5)3]2
MDL Number MFCD00044273
CAS 13938-94-8
Beilstein 4618896
Synonym Carbonylbis(triphenylphosphine)rhodium(I) chloride
Solubility Information Soluble in acetone, chloroform, ethanol
Health Hazard 1 Warning
Color Yellow
Melting Point 224° to 227°C (dec.)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Concentration Rh 14.9% min.
Quantity 0.25g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.