Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonylchlorobis(triphenylphosphine)iridium(I)

Chemical Identifiers

Chemical Name or Material Carbonylchlorobis(triphenylphosphine)iridium(I)
CAS 14871-41-1
Formula Weight 780.27
MDL Number MFCD00009590
UN Number UN2811
Synonym Vaska's catalyst; Chlorocarbonylbis(triphenylphosphine)iridium(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3941301

 
alfa aesar™
3941301
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3941303

 
alfa aesar™
3941303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3941306

 
alfa aesar™
3941306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 780.27
Chemical Name or Material Carbonylchlorobis(triphenylphosphine)iridium(I)
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00009590
CAS 14871-41-1
UN Number UN2811
Synonym Vaska's catalyst; Chlorocarbonylbis(triphenylphosphine)iridium(I)
Solubility Information Soluble in chloroform, toluene; Slightly soluble in acetone, alcohol
Health Hazard 1 Danger
Melting Point 215°C (dec.)
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 0.25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.