Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonyl cyanide 3-chlorophenylhydrazone, 98%

Chemical Identifiers

Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone
CAS 555-60-2
Molecular Formula C9H5ClN4
Formula Weight 204.62
MDL Number MFCD00001848
UN Number UN3439
Beilstein 1842102
Synonym Mesoxalonitrile 3-chlorophenylhydrazone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL06932MC

 
alfa aesar™
L06932MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL06932ME

 
alfa aesar™
L06932ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.62
Chemical Name or Material Carbonyl cyanide 3-chlorophenylhydrazone
Assay 98%
Molecular Formula C9H5ClN4
CAS 555-60-2
Synonym Mesoxalonitrile 3-chlorophenylhydrazone
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Odor Odorless
MDL Number MFCD00001848
UN Number UN3439
Beilstein 1842102
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number FG5600000
Quantity 100mg
Melting Point ≈175°C

Applications
Carbonyl cyanide 3-chlorophenylhydrazone is a widely used uncoupler of oxidative phosphorylation.

Solubility
Insoluble in water. Soluble in DMSO (5 mg/ml), ethanol (1 mg/ml) and methanol (10 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C