missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 21%

Hydroformylation

Manufacturer:  Alfa Aesar™ 1045801

 View more versions of this product

Catalog No. AA1045801



Specifications

Specifications

Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)
0.25g
MFCD00064611
Rh 21%
492.32
Yellow
25470-96-6
14,356
Soluble in acetone and chlorinated solvents
Crystals

Chemical Identifiers

25470-96-6
MFCD00064611
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents