Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I), Rh 21%

Hydroformylation

Chemical Identifiers

Chemical Name or Material Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)
CAS 25470-96-6
Formula Weight 492.32
MDL Number MFCD00064611
Merck Index 14,356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1045801

  SDS
alfa aesar™
1045801
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1045803

  SDS
alfa aesar™
1045803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1045806

  SDS
alfa aesar™
1045806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 492.32
Merck Index 14,356
TSCA TSCA
Color Yellow
Concentration Rh 21%
Quantity 0.25g
Chemical Name or Material Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)
Physical Form Crystals
CAS 25470-96-6
Solubility Information Soluble in acetone and chlorinated solvents
MDL Number MFCD00064611