Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonic Anhydrase Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67611LPL

 View more versions of this product

Catalog No. AAJ67611LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Carbonic Anhydrase Digest Standard
Quantity 1 vial
Storage Requirements -30°C to -10°C
CAS 9001-03-0
Synonym Carbonic Anhydrase; Carbonate Dehydratase
Format Lyophilized powder
Odor Odorless
Solubility Soluble in water
Description Off-white to pale brown
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.