Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbonate, 0.2M buffer soln., pH 9.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Carbonate
CAS 497-19-8
Name Note low endotoxin
MDL Number MFCD00003494
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67384AE

 
alfa aesar™
J67384AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67384AK

 
alfa aesar™
J67384AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Carbonate
CAS 497-19-8
Name Note low endotoxin
Physical Form Liquid
MDL Number MFCD00003494
pH 9.0
Concentration 0.2M buffer soln.
TSCA TSCA
Color Colorless
Quantity 100mL