Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbon yarn, woven from 0.076mm (0.003 in.) dia. fibers

Chemical Identifiers

Chemical Name or Material Carbon yarn
Name Note Woven from 0.076mm (0.003 in.) dia. fibers
MDL Number MFCD00133992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39721G5

  SDS
alfa aesar™
39721G5
5m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39721G9

  SDS
alfa aesar™
39721G9
25m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5m
Chemical Name or Material Carbon yarn
Name Note Woven from 0.076mm (0.003 in.) dia. fibers
MDL Number MFCD00133992
TSCA TSCA