missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™

Manufacturer:  LabChem LC129107

 View more versions of this product

Catalog No. LC129107



Specifications

Specifications

Carbon Standard
877-24-7,7664-38-2,877-24-7,7732-18-5
C8H5KO4
1000ppm ±10ppm Organic (1mL = 1mg C)
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
204.23
Passes Test
Poly Bottle
Traceable to NIST
125mL
1000ppm, Organic (1mL = 1mg C) (Potassium Hydrogen Phthalate)
99.49,0.3,0.21
KHC8H4O4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
Liquid
Certified
Soluble in water
Colorless

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions