missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Organic Carbon Standard, 1000 ppm C, Ricca Chemical
Supplier: Ricca Chemical Company 184725
Specifications
Organic Carbon Standard | |
Cubitainer | |
97.0,0.46,0.21 | |
Colorless | |
C8H5KO4 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
204.222 | |
1 mL = 1 mg C |
877-24-7,7664-38-2,877-24-7,7732-18-5 | |
10 L | |
100.0,0.48,0.22 | |
1000ppm | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
potassium;2-carboxybenzoate | |
23676735 | |
Liquid |
Chemical Identifiers
877-24-7 | |
204.222 | |
23676735 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
C8H5KO4 | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
potassium;2-carboxybenzoate |