Please login to your online account to display your discounted pricing

Carbon Standard (Organic), 1000ppm, Ricca Chemical


Quantity 250mL

Manufacturer: ricca chemical company  RIR1847000250C

 View more versions of this product

Catalog No. R1847000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Organic Carbon Standard
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7664-38-2
CAS Min % 0.46
CAS Max % 0.48
CAS 877-24-7
CAS Min % 0.21
CAS Max % 0.22
Physical Form ear colorless liquid
Quantity 250mL

Specifications

Chemical Name or Material Organic Carbon Standard
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7664-38-2
CAS Min % 0.46
CAS Max % 0.48
CAS 877-24-7
CAS Min % 0.21
CAS Max % 0.22
Physical Form ear colorless liquid