Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbon sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Carbon sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00133992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40830KS

  SDS
alfa aesar™
40830KS
1 ea. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 ea.
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Physical Form Hot Pressed
Assay 99.999%
Assay Percent Notes (metals basis)
Chemical Name or Material Carbon sputtering target
MDL Number MFCD00133992