Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbon sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Carbon sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00133992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40832KS

 
alfa aesar™
40832KS
1 ea. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Carbon sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Physical Form Hot Pressed
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00133992