Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbon powder, activated, Norit™ D10

Chemical Identifiers

Chemical Name or Material Carbon powder
CAS 7440-44-0
Name Note Activated, Norit™ D10
MDL Number MFCD00133992
UN Number UN1362
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH366910P

 
alfa aesar™
H366910P
2000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3669122

  SDS
alfa aesar™
H3669122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3669136

 
alfa aesar™
H3669136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Carbon powder
TSCA TSCA
Name Note Activated, Norit™ D10
Physical Form Powder
MDL Number MFCD00133992
CAS 7440-44-0
UN Number UN1362
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities
may catch fire.
Health Hazard 3 GHS P Statement
P210-P280-P235+P410-P240-P370+P378a-P420
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from su
DOT Information Transport Hazard Class: 4.2; Packaging Group: III; Proper Shipping Name: CARBON, ACTIVATED
RTECS Number FF5250100
Color Black
Quantity 2000g

Applications
Carbon powder is used as such in purifying metals, in electrodes, electrical devices and steel.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P235+P410-P240-P370+P378a-P420
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from su

GHS H Statement
H228-H252
Flammable solid.
Self-heating in large quantities
may catch fire.