Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbon felt 3.18mm thick, 99.0%, Alfa Aesar

$30.78 - $30.78

Chemical Identifiers

Chemical Name or Material Carbon felt
Name Note Processed at 1400°C
MDL Number MFCD00133992
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43199RR

 
alfa aesar™
43199RR
10 x 10cm Each for $30.78
Description & Specifications

Specifications

Quantity 10 x 10cm
Chemical Name or Material Carbon felt
EINECS Number 231-955-3
Name Note Processed at 1400°C
Assay Percent Range 99.0%
MDL Number MFCD00133992