Please login to your online account to display your discounted pricing

Carbon Disulfide, EMD Millipore

For spectroscopy Uvasol™

$1,128.29 - $1,128.29

Chemical Identifiers

Chemical Name or Material Carbon Disulfide
CAS 75-15-0
Molecular Formula CS2
Formula Weight 76.13
UN Number 1131
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1022101000

 
emd millipore corporation
1022101000
Glass Bottle 1L Each for $1,128.29
Description & Specifications

Specifications

Boiling Point 46.5°C
Melting Point −111.6°C
Formula Weight 76.13
Quantity 1L
Chemical Name or Material Carbon Disulfide
Ignition Point 95°C (203°F)
Odor Charactristic
Packaging Glass Bottle
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Vapor Pressure 398 hPa (20°C)
Flash Point −30°C (−22°F)
Physical Form Liquid
Color Colorless
Molecular Formula CS2
CAS 75-15-0
UN Number 1131
Solubility Information Soluble in water: 2.1g/L at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H315 Causes skin irritation.
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Density 1.26g/cm3 at 20°C
Health Hazard 3 H319: Causes serious eye irritation.
Safety and Handling

H225: Highly flammable liquid and vapour.

H319: Causes serious eye irritation.

H315 Causes skin irritation.