Please login to your online account to display your discounted pricing

Carbon Disulfide (Certified ACS), Fisher Chemical


Quantity 500mL
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  FLC184500

 View more versions of this product

Catalog No. C184-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Boiling Point 46°C
Melting Point −111°C
Formula Weight 76.14
Quantity 500mL
Assay Percent Range ≥99.9%
DOT Information DOT Class 3, : Flammable Liquid
Packaging Amber Glass Bottle
Flash Point 35°C (95°F)
UN Number 1120; Colorless
Physical Form Liquid
Vapor Pressure 297.5mmHg at 20°C
Purity Grade Certified ACS Reagent Grade
Viscosity 0.363 cps at 20 deg C
Color Light Yellow
Sulfur Dioxide P.T. (about 2.5ppm)
MDL Number MFCD00011321
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Chemical Name or Material Carbon Disulfide
Hydrogen Sulfide P.T. (about 1.5ppm)
Residue after Evaporation 0.002% max.
Cleanup Symbol Solvent
Water 0.05% max.
CAS 75-15-0
Synonym Carbon Bisulfide
Health Hazard 3 Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:4
Instability:0
Molecular Formula CS2

Specifications

Chemical Name or Material Carbon Disulfide
CAS 75-15-0
Purity Grade Certified ACS Reagent Grade
Assay ≥99.9%
Assay Percent Range ≥99.9%
Molecular Formula CS2
Formula Weight 76.14
MDL Number MFCD00011321
Synonym Carbon Bisulfide
Accessories
 

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:4
Instability:0

DANGER!

 

SDS & Safety Documents