missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carbon Disulfide (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLC184212
Specifications
Carbon Disulfide | |
75-15-0 | |
46°C | |
35°C (95°F) | |
Liquid | |
≥99.9% | |
MFCD00011321 | |
0.363 cps at 20 deg C | |
0.05% max. | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
methanedithione | |
6348 | |
76.14 | |
Certified ACS Reagent |
-111°C | |
Yellow | |
2.5 L | |
Glass Bottle | |
0.002% max. | |
CS2 | |
297.5mmHg at 20°C | |
1120; Colorless | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
C(=S)=S | |
76.131 | |
CHEBI:23012 | |
≥99.9% |
Chemical Identifiers
75-15-0 | |
76.131 | |
QGJOPFRUJISHPQ-UHFFFAOYSA-N | |
6348 | |
methanedithione |
CS2 | |
MFCD00011321 | |
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide | |
CHEBI:23012 | |
C(=S)=S |
Safety and Handling
DANGER!
Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .
NFPA
Health:3
Flammability:4
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid