Please login to your online account to display your discounted pricing

Carbon disulfide, 99.9%, extra pure, low in benzene content, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC177285000

 View more versions of this product

Catalog No. AC177285000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Carbon disulfide
Name Note Low in Benzene Content
CAS 75-15-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Harmful if inhaled.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
RTECS Number FF6650000
EINECS Number 200-843-6
TSCA TSCA
Purity Grade Extra Pure
Physical Form Liquid
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula CS2
Linear Formula CS2
Formula Weight 76.13
MDL Number MFCD00011321
Benzene 0.003% max.
Fieser 01,114; 05,94; 06,95
Density 1.2620g/mL
Packaging Glass bottle
Residue after Evaporation 0.002% max.
Sulfur (S) 0.0003% max. (from hydrogen sulfide) typically negative
Color Clear
Quantity 500mL

Specifications

Chemical Name or Material Carbon disulfide
Name Note Low in Benzene Content
CAS 75-15-0
Purity Grade Extra Pure
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula CS2
Linear Formula CS2
Formula Weight 76.13
MDL Number MFCD00011321
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Harmful if inhaled.

SDS & Safety Documents