Please login to your online account to display your discounted pricing

Carbon disulfide, 99.9%, extra pure, low in benzene content, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Carbon disulfide
Name Note Low in Benzene Content
CAS 75-15-0
Purity Grade Extra Pure
Assay ≥99.9%
Assay Percent Range 99.9%
Molecular Formula CS2
Linear Formula CS2
Formula Weight 76.13
MDL Number MFCD00011321
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177280250

  SDS
thermo fisher scientific
177280250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC177285000

  SDS
thermo fisher scientific
AC177285000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.9%
Physical Form Liquid
Formula Weight 76.13
Purity Grade Extra Pure
Quantity 25mL
Color Clear
EINECS Number 200-843-6
RTECS Number FF6650000
TSCA TSCA
Density 1.2620g/mL
Fieser 01,114; 05,94; 06,95
Packaging Glass bottle
Residue after Evaporation 0.002% max.
Sulfur (S) 0.0003% max. (from hydrogen sulfide) typically negative
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly fl
Health Hazard 3 GHS P Statement Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/
Benzene 0.003% max.
Chemical Name or Material Carbon disulfide
Health Hazard 1 Danger
Name Note Low in Benzene Content
Linear Formula CS2
Molecular Formula CS2
MDL Number MFCD00011321
CAS 75-15-0
Assay Percent Range 99.9%
Safety and Handling

Danger

GHS P Statement Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/

GHS P Statement Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Suspected of damaging fertility. Suspected of damaging the unborn child. Highly fl