missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbon disulfide, 99.9+%, Alfa Aesar

Manufacturer:  Alfa Aesar 32472K2

 View more versions of this product

Catalog No. AA32472K2



Specifications

Specifications

Carbon disulfide
99.9+%
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
C(=S)=S
76.131
CHEBI:23012
Liquid
-30°F (-22°F)
1L
75-15-0
CS2
QGJOPFRUJISHPQ-UHFFFAOYSA-N
methanedithione
6348
76.14
1.2632g/mol
46°C

Chemical Identifiers

75-15-0
76.131
carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
CHEBI:23012
C(=S)=S
CS2
QGJOPFRUJISHPQ-UHFFFAOYSA-N
6348
methanedithione
SDS Documents