Please login to your online account to display your discounted pricing

Carbon black, acetylene, 50% compressed, 99.9+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Carbon black, acetylene
CAS 1333-86-4
Name Note 50% compressed
MDL Number MFCD00133992
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3972430

  SDS
alfa aesar
3972430
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 250g
Chemical Name or Material Carbon black, acetylene
TSCA TSCA
Density Bulk density 80 to 120g/L
Name Note 50% compressed
Assay ≥99.9%
MDL Number MFCD00133992
CAS 1333-86-4
UN Number UN1325
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P210-P280-P281-P308+P313-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECS Number FF5800000
Odor Odorless
Color Black
Safety and Handling

Warning

GHS P Statement
P210-P280-P281-P308+P313-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.