missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbon black, acetylene, 100% compressed, >99.9%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4552730

 View more versions of this product

Catalog No. AA4552730



Specifications

Specifications

Black powder
1.8 to 2.1g/cm3 at 20°C
250g
>99.9%
MFCD00133992
12.01
Carbon black
Black
1333-86-4
C
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Powder

Chemical Identifiers

1333-86-4
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
C
MFCD00133992
Safety and Handling

Safety and Handling

H228-H351 : Flammable solid. Suspected of causing cancer.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 215-609-9

SDS Documents