Please login to your online account to display your discounted pricing

Carbon, Activated, Powder, J.T.Baker™

$228.71 - $839.46

Chemical Identifiers

Chemical Name or Material Carbon
CAS 7440-44-0
Name Note Activated
Molecular Formula C
Formula Weight 12.01
Synonym DARCO™G-60
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002803

 
avantor performance
E34309
Poly Pail 3kg Each for $839.46

02002843

 
avantor performance
E34307
Wide Mouth Amber Glass Bottle 500g Each for $228.71
Description & Specifications

Specifications

Formula Weight 12.01
Chemical Name or Material Carbon
CAS 7440-44-0
Synonym DARCO™G-60
Packaging Poly Pail
Physical Form Powder
Quantity 3kg
Name Note Activated
Molecular Formula C