Please login to your online account to display your discounted pricing

Carbon, activated, NorIT™ CN 1, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404045000

 View more versions of this product

Catalog No. AC404045000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 12
Quantity 500g
Color Black
Merck Index 15, 1807
TSCA TSCA
EINECS Number 231-153-3
RTECS Number FF5250100
pH 5.5 to 8
Molecular Formula C
Phosphate 3.5% max.
Calcium (Ca) 200ppm max.
Additional Information methylene blue adsorption (g/100g):> 25
Loss on Drying 15% max.
MDL Number MFCD00133992
Chemical Name or Material Activated charcoal, NORIT™ CN 1
Name Note NORIT CN 1
CAS 7440-44-0
Iron (Fe) 150ppm max.
Packaging Glass bottle
Trace Analysis trace analysis by acid extraction
Synonym Carbon, activated

Specifications

Chemical Name or Material Activated charcoal, NORIT™ CN 1
Name Note NORIT CN 1
CAS 7440-44-0
Molecular Formula C
Formula Weight 12
MDL Number MFCD00133992
Synonym Carbon, activated