missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Activated charcoal, NORIT™ CN 1, Acros Organics™

$62.20 - $62.20

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC404045000 SDS View Documents Acros Organics
404045000
500g Glass bottle Each for $62.20
 
Description

Description

Filter aid
Specifications

Chemical Identifiers

7440-44-0
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

15% max.
Black
500g
7440-44-0
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12
Activated charcoal, NORIT™ CN 1, NORIT CN 1
Glass bottle
3652.0°C
5.5 to 8
C
15, 1807
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents
[C]
12.011
CHEBI:27594
Powder
Safety and Handling

Safety and Handling

EINECSNumber : 231-153-3

RTECSNumber : FF5250100

TSCA : TSCA