missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Carbon, activated, -20+40 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045478A3
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Carbon | |
Black | |
-20+40 Mesh | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.011 | |
CHEBI:27594 |
7440-44-0 | |
2.26g/cm3 at 20°C | |
C | |
2 kg | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
Powder |
Chemical Identifiers
7440-44-0 | |
12.011 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Safety and Handling
H500
EINECSNumber : 231-153-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only