missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbol Fuchsin, pure, ACROS Organics™

$77.12 - $234.06

Chemical Identifiers

CAS 4197-24-4
Molecular Formula C21H22ClN3
Molecular Weight (g/mol) 351.878
MDL Number MFCD00143923
InChI Key HZLHRDBTVSZCBS-GHTYLULLSA-N
PubChem CID 91997463
IUPAC Name 4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride
SMILES [H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC195790050 SDS View Documents Acros Organics
195790050
5g Glass bottle Each for $77.12
 
AC195790250 SDS View Documents Acros Organics
195790250
25g Glass bottle Each for $234.06
 
AC195791000 SDS View Documents Acros Organics
AC195791000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

4197-24-4
351.878
HZLHRDBTVSZCBS-GHTYLULLSA-N
4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride
C21H22ClN3
MFCD00143923
91997463
[H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]

Specifications

4197-24-4
MFCD00143923
Authentic
4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride
(in water) about 542nm
Crystalline Powder
Glass bottle
Complies
5g
C21H22ClN3
HZLHRDBTVSZCBS-GHTYLULLSA-N
[H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]
351.878
91997463
Pure
(for microscopy) passes
Green
Carbol Fuchsin, A mixture of Basic Fuchsin and phenol.
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing genetic defects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical attention/advice.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

DOTInformation : DOT Class 6.1

EINECSNumber : 224-086-6