Please login to your online account to display your discounted pricing

Carbic anhydride, 99+%, ACROS Organics™

$61.64 - $304.38

Chemical Identifiers

Chemical Name or Material Carbic anhydride
CAS 129-64-6
Assay ≥99.0% (anhydride), balance: himic acid
Assay Percent Range 99+%
Molecular Formula C9H8O3
Formula Weight 164.16
MDL Number MFCD00151106
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC134511000

  SDS
thermo fisher scientific
AC134511000
Glass bottle 100g Each for $61.64

AC134510010

  SDS
thermo fisher scientific
134510010
Glass bottle 1kg Each for $304.38
Description & Specifications

Specifications

Assay ≥99.0% (anhydride), balance: himic acid
Physical Form Crystalline Powder
Formula Weight 164.16
Quantity 100g
Color White
Density 1.0800g/mL
Melting Point 162° to 165°C
Merck Index 15, 1797
Chemical Name or Material Carbic anhydride
Molecular Formula C9H8O3
Infrared Spectrum Authentic
MDL Number MFCD00151106
Packaging Glass bottle
Synonym endo-cis-5-Norbornene-2, 3-dicarboxylic anhydride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99+%
CAS 129-64-6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.