Please login to your online account to display your discounted pricing

Carbenicillin (Disodium Salt), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26485

 View more versions of this product

Catalog No. BP26485

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 422.4
Boiling Point 738°C
Melting Point 200°C
Quantity 5g
Physical Form Solid
Molecular Formula C17H16N2Na2O6S
Water 6% Max.
Color Off-white
Identification Pass Test
Name Note Disodium Salt
Assay Percent Range ≥92 %
ChemAlert Storage Symbol Gray
Packaging Amber Glass
Potency 770μg/mg min.
Health Hazard 2 WARNING!
Recommended Storage 0° to 5°C
Chemical Name or Material Carbenicillin
CAS 4800-94-6
Synonym α-Carboxybenzylpenicillin
Purity 92% min.
Health Hazard 3 Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0

Specifications

Chemical Name or Material Carbenicillin
Name Note Disodium Salt
CAS 4800-94-6
Physical Form Solid
Assay Percent Range ≥92 %
Molecular Formula C17H16N2Na2O6S
Formula Weight 422.4
Synonym α-Carboxybenzylpenicillin
Accessories
 

Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

0° to 5°C

SDS & Safety Documents