Learn More
Carbenicillin disodium salt, Thermo Scientific Chemicals
Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities.
$160.32 - $2021.37
Chemical Identifiers
CAS | 4800-94-6 |
---|---|
Molecular Formula | C17H16N2Na2O6S |
Molecular Weight (g/mol) | 422.36 |
MDL Number | MFCD00077683 |
InChI Key | RTYJTGSCYUUYAL-YCAHSCEMSA-L |
PubChem CID | 20933 |
ChEBI | CHEBI:34609 |
SMILES | [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6194903
|
Thermo Scientific Chemicals
J6194903 |
1 g |
Each for $160.32
|
|
|||||
AAJ6194906
|
Thermo Scientific Chemicals
J6194906 |
5 g |
Each for $472.18
|
|
|||||
AAJ6194914
|
Thermo Scientific Chemicals
J6194914 |
25 g |
Each for $2,021.37
|
|
|||||
Description
Carbenicillin disodium salt is a water-soluble antibiotic which is effective against gram-negative bacteria.Carbenicillin disodium salt is widely utilized as an antibiotic which is used to control bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It has been involved in the study of the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
- In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium
Application
- Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
- It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
Chemical Identifiers
4800-94-6 | |
422.36 | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
CHEBI:34609 |
C17H16N2Na2O6S | |
MFCD00077683 | |
20933 | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Specifications
Carbenicillin Disodium Salt | |
4800-94-6 | |
MFCD00077683 | |
14,1792 | |
RTYJTGSCYUUYAL-YCAHSCEMSA-L | |
disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
20933 | |
422.36 | |
1 g |
Powder | |
C17H16N2Na2O6S | |
5722128 | |
Soluble in water and alcohol. | |
[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
422.36 | |
CHEBI:34609 | |
White | |
Carbenicillin disodium salt |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c
H317-H334-H335
EINECSNumber : 225-360-8
RTECSNumber : ON9105000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only