missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Carbazole-2-boronic acid pinacol ester, tech. 90%

Manufacturer:  Alfa Aesar™ H55484MD

 View more versions of this product

Catalog No. AAH55484MD



Specifications

Specifications

Carbazole-2-boronic acid pinacol ester
871125-67-6
MFCD07784368
RLSJGSFDSSYNPL-UHFFFAOYSA-N
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
46738013
Solid
Technical
250mg
C18H20BNO2
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
293.173
293.17
90%

Chemical Identifiers

871125-67-6
293.173
RLSJGSFDSSYNPL-UHFFFAOYSA-N
46738013
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
C18H20BNO2
MFCD07784368
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents