Please login to your online account to display your discounted pricing

Carbazeran citrate, Tocris™

Aldehyde oxidase substrate; PDE inhibitor

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Carbazeran citrate
Assay Percent Range >98%
Molecular Formula C18H24N4O4·C6H8O7
Formula Weight 552.53
Synonym Ethyl 1-(6, 7-dimethoxy-1-phthalazinyl)-4-piperidinyl carbamate citrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

442050

 
r & d systems
RDS442050
50mg Each for $546.19

442010

 
r & d systems
RDS442010
10mg Each for $129.05
Description & Specifications

Specifications

Formula Weight 552.53
Quantity 50mg
Chemical Name or Material Carbazeran citrate
Assay Percent Range >98%
Molecular Formula C18H24N4O4·C6H8O7
CAS 153473-94-0
Synonym Ethyl 1-(6, 7-dimethoxy-1-phthalazinyl)-4-piperidinyl carbamate citrate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C