Please login to your online account to display your discounted pricing

Carbamazepine, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  15354905

 View more versions of this product

Catalog No. ICN15354905

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 236.27
Color White
Quantity 5g
EINECS Number 206-062-7
CAS 298-46-4
Molecular Formula C15H12N2O
Synonym 5H-Dibenz[b, f]azepine-5-carboxamide
MDL Number MFCD00005073
Chemical Name or Material Carbamazepine
Recommended Storage Store at Room Temperature (15 to 30°C).

Specifications

Chemical Name or Material Carbamazepine
CAS 298-46-4
Physical Form Powder
Molecular Formula C15H12N2O
Formula Weight 236.27
MDL Number MFCD00005073
Synonym 5H-Dibenz[b, f]azepine-5-carboxamide
Accessories

Store at Room Temperature (15 to 30°C).