Please login to your online account to display your discounted pricing

Carbamazepine, MP Biomedicals™

$94.95 - $94.95

Chemical Identifiers

Chemical Name or Material Carbamazepine
CAS 298-46-4
Physical Form Powder
Molecular Formula C15H12N2O
Formula Weight 236.27
MDL Number MFCD00005073
Synonym 5H-Dibenz[b, f]azepine-5-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15354905

 
mp biomedicals
15354905
5g Each for $94.95
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 236.27
Color White
Quantity 5g
EINECS Number 206-062-7
Molecular Formula C15H12N2O
CAS 298-46-4
Chemical Name or Material Carbamazepine
MDL Number MFCD00005073
Synonym 5H-Dibenz[b, f]azepine-5-carboxamide
Recommended Storage Store at Room Temperature (15 to 30°C).
Safety and Handling

Store at Room Temperature (15 to 30°C).