missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Carbadox, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6028422

 View more versions of this product

Catalog No. AAJ6028422



Description

Description

Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbadox has been used to suppress aldosterone production, to study the spread of shiga toxin-producing Escherichia coli (STEC) strains, and to compare different antimicrobial activities. Carbadox is a quinoxaline-di-N-oxide antibiotic that is generally used in feed to swine to prevent dysentery and improve feed efficiency. It inhibits bacteria by intercalation and induction of mutations in bacterial genome.

Solubility
Soluble in 1N NaOH (50 mg/ml), and water (partly).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Specifications

Carbadox
239°C to 240°C
18°C (64°F)
MFCD00057293
UN1325
carbadox, unii-m2x04r2e2y, getroxel, mecadox, dsstox_cid_23913, dsstox_rid_80088, dsstox_gsid_43913, fortigro, karbadox czech, methyl n-e-1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene methyl iminocarbamate
BPMVRAQIQQEBLN-UHFFFAOYSA-N
methyl N-[(E)-(1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene)methyl]iminocarbamate
5353472
≥98%
6804-07-5
Yellow
C11H10N4O4
100 g
14,1780
Soluble in 1N NaOH (50mg/ml),and water (partly).
COC(=O)N=NC=C1C=[N+](C2=CC=CC=C2N1O)[O-]
262.23
262.22
Solid

Chemical Identifiers

6804-07-5
MFCD00057293
Safety and Handling

Safety and Handling

GHS H Statement
H228-H350-H302
Flammable solid.
May cause cancer.
Harmful if swallowed.

P201-P202-P210-P240-P241-P264b-P270-P281-P301+P312-P308+P313-P330-P370+P378q-P501c

H228-H302-H350

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : FE2779000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only