missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Erlab™ CaptairStore 822 B V2 Filtering Chemical Storage Cabinet with Pull Drawer

Filtering Chemical Storage Cabinets designed with advanced carbon filtration to keep vapors at bay and also provide storage under bench.

Supplier:  Erlab™ 822FSCV2

Catalog No. 08444048



Description

Description

Erlab™'s CaptairStore Filtering Chemical Storage Cabinets are designed with advanced carbon filtration, Flex Technology and a simple, innovative way to keep you safe. With the push of one button, it will set you free of limitations in order to focus on what is most important: your work.

  • Cabinet is filtered 24 hours, 7 days a week
  • No build up of vapors or odors present within the cabinet as air is continuously turned over
  • No HVAC connection required to the outside
  • Units arrive completely knocked down and require assembly (with exception of 822 B)
  • Includes one storage compartment with large volume drawer
Specifications

Specifications

Lifetime
22.8 in.
29.7 in.
26 in.
580 mm
652 mm
Captair Ductless Filtering Fume Hoods
1
33 lb.
Stainless Steel (304)
1
755 mm
Storage Cabinet, Power Cord, Fan Motor, Spill Mat, Sliding Shelf
Extractable
15 kg
1
Chemical Storage
AFNOR NF X 15-211:2009
White
SDS
Documents

Documents

Promotions

Promotions