missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CAPSO sodium salt, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444131000

 View more versions of this product

Catalog No. AC444131000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Biological buffer
Specifications

Specifications

0.050 max. (0.1M H2O) at 280nm
CAPSO sodium salt
Glass bottle
White
100g
102601-34-3
C9H18NNaO4S
(10% in water) Clear colorless
C1CCC(CC1)NCC(CS(=O)(=O)[O-])O.[Na+]
259.3
259.3
98%
(10% solution in water) clear, colorless
Authentic
5% Max.
206°C to 211°C
10.9 to 11.6 (1% aq. soln. at 25°C)
98%
3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso sodium salt, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate
GGPORGHJKBOGDV-UHFFFAOYSA-M
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
25000353
Crystalline Powder

Chemical Identifiers

102601-34-3
259.3
3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso sodium salt, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
C9H18NNaO4S
GGPORGHJKBOGDV-UHFFFAOYSA-M
25000353
C1CCC(CC1)NCC(CS(=O)(=O)[O-])O.[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

SDS Documents