missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPSO, 0.2M buffer soln., pH 9.5

$83.27 - $130.46

Chemical Identifiers

CAS 102601-34-3
Molecular Formula C9H18NNaO4S
Molecular Weight (g/mol) 259.296
MDL Number MFCD00070063
InChI Key GGPORGHJKBOGDV-UHFFFAOYSA-M
PubChem CID 25000353
IUPAC Name sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
SMILES C1CCC(CC1)NCC(CS(=O)(=O)[O-])O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63242AE View Documents Alfa Aesar™
J63242AE
100mL Each for $83.27
 
AAJ63242AK View Documents Alfa Aesar™
J63242AK
250mL Each for $130.46
 
Specifications

Chemical Identifiers

102601-34-3
259.296
GGPORGHJKBOGDV-UHFFFAOYSA-M
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
C9H18NNaO4S
MFCD00070063
25000353
C1CCC(CC1)NCC(CS(=O)(=O)[O-])O.[Na+]

Specifications

102601-34-3
MFCD00070063
GGPORGHJKBOGDV-UHFFFAOYSA-M
sodium;3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate
25000353
Odorless
100mL
C9H18NNaO4S
0.2M buffer soln.
C1CCC(CC1)NCC(CS(=O)(=O)[O-])O.[Na+]
259.296
Liquid
9.5
CAPSO buffer soln.