Please login to your online account to display your discounted pricing

Capsazepine, Tocris


Vanilloid receptor antagonist; Also activator of ENaCδ

Quantity 10mg

Manufacturer: r & d systems  046410

 View more versions of this product

Catalog No. 046410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 376.9
Assay Percent Range >99%
Chemical Name or Material Capsazepine
Molecular Formula C19H21ClN2O2S
CAS 138977-28-3
Synonym N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Capsazepine
Assay Percent Range >99%
Molecular Formula C19H21ClN2O2S
Formula Weight 376.9
Synonym N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide
Accessories

Store at Room Temperature