Please login to your online account to display your discounted pricing

CAPS (Fine White Crystals), Fisher BioReagents


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLBP321500

 View more versions of this product

Catalog No. BP321-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 221.32
Melting Point 324°C
Physical Form Powder/Solid
Assay ≥98%
Quantity 500g
Merck Index 15, 1769
Molecular Formula C9H19NO3S
pH 3.0 to 7.0
Color White
Identification Pass Test
Health Hazard 3 Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:0
Linear Formula C6H11NH(CH2)3SO3H
Recommended Storage RT
CAS 1135-40-6
Additional Information Vapor Pressure: Negligible
Packaging Glass Bottle
pKa 10.2 to 10.6 (at 25°C)
Synonym 3-Cyclohexylamino-1-propanesulfonic Acid
Assay Percent Range ≥98 %
Health Hazard 2 WARNING!
Chemical Name or Material CAPS
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material CAPS
CAS 1135-40-6
Physical Form Powder/Solid
Assay ≥98%
Assay Percent Range ≥98 %
Molecular Formula C9H19NO3S
Linear Formula C6H11NH(CH2)3SO3H
Formula Weight 221.32
Synonym 3-Cyclohexylamino-1-propanesulfonic Acid
Accessories
 

Emergency Overview
Harmful if swallowed. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

RT

SDS & Safety Documents