missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPS, 0.5M buffer soln., pH 9.5

$82.35 - $128.92

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.315
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES C1CCC(CC1)NCCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60776AE View Documents Alfa Aesar™
J60776AE
100mL Each for $82.35
 
AAJ60776AK View Documents Alfa Aesar™
J60776AK
250mL Each for $128.92
 
Specifications

Chemical Identifiers

1135-40-6
221.315
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
C9H19NO3S
MFCD00003837
70815
C1CCC(CC1)NCCCS(=O)(=O)O

Specifications

1135-40-6
MFCD00003837
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
70815
9.5
CAPS buffer soln.
C9H19NO3S
0.5M buffer soln.
C1CCC(CC1)NCCCS(=O)(=O)O
221.315
Liquid
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA