Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPS, 0.2M buffer soln., pH 9.0, low endotoxin

Chemical Identifiers

Chemical Name or Material CAPS
CAS 1135-40-6
Name Note low endotoxin
MDL Number MFCD00003837
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67007AE

  SDS
alfa aesar™
J67007AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67007AK

  SDS
alfa aesar™
J67007AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 1135-40-6
MDL Number MFCD00003837
TSCA TSCA
Quantity 100mL
Name Note low endotoxin
Physical Form Liquid
Storage Note 1 2° to 8°C
pH 9.0
Concentration 0.2M buffer soln.
Color Colorless
Chemical Name or Material CAPS
Safety and Handling

2° to 8°C