Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPS, 0.2M buffer soln., pH 9.0, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67007AK

 View more versions of this product

Catalog No. AAJ67007AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material CAPS
pH 9.0
Color Colorless
TSCA TSCA
Physical Form Liquid
Concentration 0.2M buffer soln.
MDL Number MFCD00003837
Quantity 250mL
Name Note low endotoxin
CAS 1135-40-6
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material CAPS
CAS 1135-40-6
Name Note low endotoxin
MDL Number MFCD00003837
Accessories

2° to 8°C

SDS & Safety Documents