missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPS, 0.2M buffer soln., pH 10.0, low endotoxin

$71.92 - $119.35

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.315
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES C1CCC(CC1)NCCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67292AE View Documents Alfa Aesar™
J67292AE
100mL Each for $71.92
 
AAJ67292AK View Documents Alfa Aesar™
J67292AK
250mL Each for $119.35
 
Specifications

Chemical Identifiers

1135-40-6
221.315
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
C9H19NO3S
MFCD00003837
70815
C1CCC(CC1)NCCCS(=O)(=O)O

Specifications

1135-40-6
MFCD00003837
PJWWRFATQTVXHA-UHFFFAOYSA-N
3-(cyclohexylamino)propane-1-sulfonic acid
70815
10
100mL
C9H19NO3S
0.2M buffer soln.
C1CCC(CC1)NCCCS(=O)(=O)O
221.315
Liquid
Colorless
CAPS, low endotoxin
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H335

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold