Please login to your online account to display your discounted pricing

Alfa Aesar™ CAPS, 0.2M buffer soln., pH 10.0, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67292AK

 View more versions of this product

Catalog No. AAJ67292AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material CAPS
CAS 1135-40-6
MDL Number MFCD00003837
Physical Form Liquid
TSCA TSCA
Color Colorless
Storage Note 1 2° to 8°C
pH 10
Concentration 0.2M buffer soln.
Quantity 250mL
Name Note low endotoxin

Specifications

Chemical Name or Material CAPS
CAS 1135-40-6
Name Note low endotoxin
MDL Number MFCD00003837
Accessories

2° to 8°C