missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Capitol Vial 13mL Flip-Top Vials

Thermo Scientific™ Capitol Vial 13mL Flip-Top Vials are ideal for tissue sample transport between the physician's office and laboratory.

$297.70 - $904.45

Specifications

Capacity 0.44 oz., 13mL
Capacity (Metric) 13mL
Capacity (English) 0.44 oz.
Material High-clarity resin
Sterile Yes
View More Specs

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.



Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
14-377-827 Thermo Scientific™
VETHC1500
Flip-Top Vials Case of 1500 Each for $904.45
14-377-826 Thermo Scientific™
VETHC500
Flip-Top Vials Case of 500 Each for $297.70
14-377-828 Thermo Scientific™
VETLSCL500
Flip-Top Vials w/Lock seal Case of 500 Each for $332.91
Specifications

Description

Thermo Scientific™ Capitol Vial 13mL Flip-Top Vials are ideal for tissue sample transport between physician's office and laboratory.

  • Made of high-clarity resin
  • Store and transport small body fluid samples
  • Handy pocket size vial for vitamins or pills
  • Sterile

Recommended for:

Mastitis testing and veterinary specimen collection

Warranty: 90 days

Description & Specifications

Specifications

0.44 oz., 13mL
0.44 oz.
Yes
13mL
High-clarity resin